Learn More
Description
- Human kidney tissue membrane protein lysate was prepared by isolating the membrane protein from whole tissue homogenates using a proprietary technique
- The human kidney tissue was frozen in liquid nitrogen immediately after excision and then stored at -70°C
- The membrane protein is provided in a buffer including HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
- For quality control purposes, the isolated kidney tissue membrane protein pattern on SDS-PAGE gel is shown to be consistent for each lot by visualization with coomassie blue staining
- The isolated kidney tissue membrane protein is then Western analyzed by either GAPDH or β-actin antibody to confirm there is no signal or very weak signal.
Specifications
Specifications
| Quantity | 0.1 mg |
| Tissue Condition | Tumor |
| Host Species | Human |
| Concentration | 2 mg/mL |
| Protein State | Denatured |
| Tissue | Kidney |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
