Learn More
Amsbio LLC Human Kidney (renal cell) cancer and normal adjacent tissue FFPE block
Supplier:  Amsbio LLC AMS25006
Encompass_Preferred
Human FFPE tissue blocks are processed from diseased tissues or clinically-unremarkable donor normal human tissue samples. Tissue is prepared by a certified medical pathologist, fixed in 10% neutral formalin for 24 hours and embedded in IHC-grade paraffin. The target FFPE specimen size is 0.5 x 0.5 x 0.5 cm, but it can vary significantly depending on the organ.
Catalog No. 50-204-1361
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.