Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KIR2DL1/KIR2DS5 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4715777
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1844A025 25 Tests
FAB1844A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1844A025 Supplier R&D Systems Supplier No. FAB1844A025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

KIR2DL1/KIR2DS5 Monoclonal specifically detects KIR2DL1/KIR2DS5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DL1/KIR2DS5
Applications Flow Cytometry
Classification Monoclonal
Clone 143211
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P43626
Gene Alias CD158a, CD158g, NKAT9
Gene Symbols KIR2DL1
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DL1/KIR2DS5 Accession # P43626
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3802
Test Specificity Detects human KIR2DL1/KIR2DS5 on transfected cells. It does not stain cells transfected with KIR2DL2, 2DL3, 2DL4, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.