Learn More
ACROBiosystems Human KIR3DL3 25ug
Supplier:  ACROBiosystems KI3H82F325UG
Encompass_Preferred
Biotinylated Human KIR3DL3 / CD158z Protein FcAvitag (KI3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Leu 322 (Accession # Q8N743-1).
Catalog No. 50-253-4543
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.