Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KIRREL3 ELISA Kit

Catalog No. EH288RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH288RBX10 10 x 96 Tests
EH288RBX5 5 x 96 Tests
EH288RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH288RBX10 Supplier Invitrogen™ Supplier No. EH288RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human KIRREL3 quantitates human KIRREL3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human KIRREL3.

The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. The protein encoded by this gene is a synaptic cell adhesion molecule with multiple extracellular immunoglobulin-like domains and a cytoplasmic PDZ domain-binding motif. Mutations in this gene are associated with several neurological and cognitive disorders. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 1500010O20Rik; 2900036G11Rik; KIAA1867; kin of IRRE like 3 (Drosophila); Kin of irregular chiasm-like protein 3; kin of IRRE-like protein 3; KIRRE; KIRREL3; membrane protein mKirre; mKIAA1867; mKirre; MRD4; NEPH2; nephrin-like 2; nephrin-like protein 2; PRO4502; Processed kin of IRRE-like protein 3; SST4; UNQ5923/PRO4502/PRO19814; x kin of IRRE like 3
Assay Range 0.21 to 50 ng/mL
Assay Sensitivity 0.21 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 84623
Gene Symbol KIRREL3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target KIRREL3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.