Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KIRREL3 Full-length ORF (NP_115920.1, 1 a.a. - 778 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89949932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-949-932 10 μg
89-949-933 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-949-932 Supplier Abnova™ Supplier No. H00084623P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the nephrin-like protein family. These proteins are expressed in fetal and adult brain, and also in podocytes of kidney glomeruli. The cytoplasmic domains of these proteins interact with the C-terminus of podocin, also expressed in the podocytes, cells involved in ensuring size- and charge-selective ultrafiltration. Mutations in this gene are associated with mental retardation autosomal dominant type 4 (MRD4). Alternatively spliced transcript variants encoding different isoforms have been found for this gene

Sequence: MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHV

Specifications

Accession Number NP_115920.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84623
Molecular Weight (g/mol) 111.7kDa
Name KIRREL3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MKPFQLDLLFVCFFLFSQELGLQKRGCCLVLGYMAKDKFRRMNEGQVYSFSQQPQDQVVVSGQPVTLLCAIPEYDGFVLWIKDGLALGVGRDLSSYPQYLVVGNHLSGEHHLKILRAELQDDAVYECQAIQAAIRSRPARLTVLVPPDDPVILGGPVISLRAGDPLNLTCHADNAKPAASIIWLRKGEVINGATYSKTLLRDGKRESIVSTLFISPGDVENGQSIVCRATNKAIPGGKETSVTIDIQHPPLVNLSVEPQPVLEDNVVTFHCSAKANPAVTQYRWAKRGQIIKEASGEVYRTTVDYTYFSEPVSCEVTNALGSTNLSRTVDVYFGPRMTTEPQSLLVDLGSDAIFSCAWTGNPSLTIVWMKRGSGVVLSNEKTLTLKSVRQEDAGKYVCRAVVPRVGAGEREVTLTVNGPPIISSTQTQHALHGEKGQIKCFIRSTPPPDRIAWSWKENVLESGTSGRYTVETISTEEGVISTLTISNIVRADFQTIYNCTAWNSFGSDTEIIRLKEQGSEMKSGAGLEAESVPMAVIIGVAVGAGVAFLVLMATIVAFCCARSQRNLKGVVSAKNDIRVEIVHKEPASGREGEEHSTIKQLMMDRGEFQQDSVLKQLEVLKEEEKEFQNLKDPTNGYYSVNTFKEHHSTPTISLSSCQPDLRPAGKQRVPTGMSFTNIYSTLSGQGRLYDYGQRFVLGMGSSSIELCEREFQRGSLSDSSSFLDTQCDSSVSSSGKQDGYVQFDKASKASASSSHHSQSSSQNSDPSRPLQRRMQTHV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA1867/KIRRE/MGC126824/MGC126850/NEPH2/PRO4502
Common Name KIRREL3
Gene Symbol KIRREL3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.