Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human KLF4 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. IC3640A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC3640A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC3640A Supplier R&D Systems Supplier No. IC3640A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

KLF4 Polyclonal specifically detects KLF4 in Human samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining).

Specifications

Antigen KLF4
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Gene Accession No. AAH29923
Gene Alias Epithelial zinc finger protein EZF, EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Gene Symbols KLF4
Host Species Goat
Immunogen E. coli-derived recombinant human KLF4 Ala2-Phe470 Accession # AAH29923
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9314
Test Specificity Detects human KLF4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 50% cross-reactivity with recombinant mouse (rm) KLF4 and less than 1% cross-reactivity with recombinant human (rh) KLF5, rhKLF6, and rmKLF15.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.