Learn More
Gibco™ Human KLF4-TAT Recombinant Protein, PeproTech®
Description
110-08-1MG will be provided as 10 x 100 μg (110-08-100UG). 110-08-250UG will be provided as multiple 25 μg (110-08-25UG) and 100 μg (110-08-100UG) vials. 110-08-500UG will be provided as 5 x 100 μg (110-08-100UG). Recombinant Human KLF4-TAT is a 483 amino acid protein, including a 13-residue C-terminal TAT peptide, with a calculated molecular weight of 51.7 kDa. PeproTech's Recombinant Human KLF4-TAT is a mixture of the expected sequence beginning at Met1 and a truncated isoform beginning at Tyr54. Due to post-translational modifications, SDS-PAGE gel shows bands at approximately 72 and 66kDa, under reduced conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | O43474 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 9314 |
| Molecular Weight (g/mol) | 51.7 kDa |
| Name | Human KLF4-TAT |
| Quantity | 25 μg |
| Source | HEK293 cells |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.