Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KLHL21 Full-length ORF (BAG52104.1, 1 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-048 25 μg
89-950-047 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-048 Supplier Abnova™ Supplier No. H00009903P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sequence: MERPAPLAVLPFSDPAHALSLLRGLSQLRAERKFLDVTLEAAGGRDFPAHRAVLAAASPYFRAMFAGQLRESRAERVRLHGVPPDMLQLLLDFSYTGRVAVSGDNAEPLLRAADLLQFPAVKEACGAFLQQQLDLANCLDMQDFAEAFSCSGLASAAQRFILRHVGELGAEQLERLPLARLLRYLRDDGLCVPKEEAAYQLALRWVRADPPRRAAHWPQLLEAVRLPFVRRFYLLAHVEAEPLVARCPPCLRLLREARDFQAARYDRHDRGPCPRMRPRPSTGLAEILVLVGGCDQDCDELVTVDCYNPQTGQWRYLAEFPDHLGGGYSIVALGNDIYVTGGSDGSRLYDCVWRYNSSVNEWAEVAPMLKAREYHSSSVPDGLLYVVAADSTERYDHTTDSWEALQPMTYPMDNCSTTACRGRLYAIGSLAGKETMVMQCYDPDTDLWSLVDCGQLPPWSFAPKTATLNGLMYFVRDDSAEVDVYNPTRNEWDKIPSMNQVHVGGSLAVLGGKLYVSGGYDNTFELSDVVEAYDPETRAWSVVGRLPEPTFWHGSVSIFRQFMPQTFSGGRGFELDSGSDDMDPGRPRPPRDPDELH

Specifications

Accession Number BAG52104.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9903
Molecular Weight (g/mol) 93kDa
Name KLHL21 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Immunogen MERPAPLAVLPFSDPAHALSLLRGLSQLRAERKFLDVTLEAAGGRDFPAHRAVLAAASPYFRAMFAGQLRESRAERVRLHGVPPDMLQLLLDFSYTGRVAVSGDNAEPLLRAADLLQFPAVKEACGAFLQQQLDLANCLDMQDFAEAFSCSGLASAAQRFILRHVGELGAEQLERLPLARLLRYLRDDGLCVPKEEAAYQLALRWVRADPPRRAAHWPQLLEAVRLPFVRRFYLLAHVEAEPLVARCPPCLRLLREARDFQAARYDRHDRGPCPRMRPRPSTGLAEILVLVGGCDQDCDELVTVDCYNPQTGQWRYLAEFPDHLGGGYSIVALGNDIYVTGGSDGSRLYDCVWRYNSSVNEWAEVAPMLKAREYHSSSVPDGLLYVVAADSTERYDHTTDSWEALQPMTYPMDNCSTTACRGRLYAIGSLAGKETMVMQCYDPDTDLWSLVDCGQLPPWSFAPKTATLNGLMYFVRDDSAEVDVYNPTRNEWDKIPSMNQVHVGGSLAVLGGKLYVSGGYDNTFELSDVVEAYDPETRAWSVVGRLPEPTFWHGSVSIFRQFMPQTFSGGRGFELDSGSDDMDPGRPRPPRDPDELH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KIAA0469/MGC99635
Common Name KLHL21
Gene Symbol KLHL21
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.