Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Klotho Recombinant Protein, PeproTech®

Catalog No. 1005301M
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-053-01M 2 x 500 μg
10053100UG 100 μg
100-53-20UG 20 μg
10053250UG 250 μg
10053500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-053-01M Supplier Gibco™ Supplier No. 1005301M
Add to Cart
Add to Cart

Recombinant Protein

100-53-1MG will be provided as 2 x 500 μg (100-53-500UG). Recombinant Human Klotho is a glycoprotein of 516 amino acid residues that migrates at an apparent molecular weight of 65-70 kDa by SDS-PAGE analysis under reducing conditions. Recombinant Human Klotho has a calculated molecular weight of 58.6 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

KLOTHO is the systemic anti-aging hormone within the glycosidase1 superfamily. It encodes a type I membrane protein that is abundant in the kidney and brain. In mice, a deficiency in KLOTHO expression leads to various systemic phenotypes resembling human aging such as arteriosclerosis, osteoporosis, and skin atrophy together with growth retardation, short life-span and infertility. Transgenic mice overexpressing KLOTHO have an extended life span by inhibiting insulin/IGF1 signaling. KLOTHO is involved in the regulation of calcium/phosphorus homeostasis by inhibiting the synthesis of active vitamin D and identified as a potential tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UEF7
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 9365
Molecular Weight (g/mol) 58.6 kDa
Name Human Klotho
Purification Method Purified
Quantity 2 x 500 μg
Source CHO cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein
Common Name Klotho
Gene Symbol KL
Biological Activity Determined by the dose-dependent stimulation of the proliferation of murine NIH-3T3 cells. Recombinant Human Klotho is effective in a concentration range of 0.5-2.0 ug/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence EPGDGAQTWA RFSRPPAPEA AGLFQGTFPD GFLWAVGSAA YQTEGGWQQH GKGASIWDTF THHPLAPPGD SRNASLPLGA PSPLQPATGD VASDSYNNVF RDTEALRELG VTHYRFSISW ARVLPNGSAG VPNREGLRYY RRLLERLREL GVQPVVTLYH WDLPQRLQDA YGGWANRALA DHFRDYAELC FRHFGGQVKY WITIDNPYVV AWHGYATGRL APGIRGSPRL GYLVAHNLLL AHAKVWHLYN TSFRPTQGGQ VSIALSSHWI NPRRMTDHSI KECQKSLDFV LGWFAKPVFI DGDYPESMKN NLSSILPDFT ESEKKFIKGT ADFFALCFGP TLSFQLLDPH MKFRQLESPN LRQLLSWIDL EFNHPQIFIV ENGWFVSGTT KRDDAKYMYY LKKFIMETLK AIKLDGVDVI GYTAWSLMDG FEWHRGYSIR RGLFYVDFLS QDKMLLPKSS ALFYQKLIEK NGFPPLPENQ PLEGTFPCDF AWGVVDNYIQ VSQLTKPISS LTKPYH
Content And Storage -20°C
Activity Determined by the dose-dependent stimulation of the proliferation of murine NIH-3T3 cells. Recombinant Human Klotho is effective in a concentration range of 0.5-2.0 ug/ml.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.