Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human KLRA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569427 Supplier Invitrogen™ Supplier No. PA569427
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 90%.

Ly49A is a gene that encodes a protein expressed on the surface of natural killer cells, which is involved in recognizing and eliminating virally infected or cancerous cells. It is located on chromosome 6 and is part of a larger family of genes that encode for similar receptors. The Ly49A protein interacts with a variety of ligands, including certain viruses and tumor cells, and is involved in regulating the activation and function of natural killer cells. Mutations or variations in the Ly49A gene have been associated with various diseases and conditions, including autoimmune disorders and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human KLRA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KLRA1P
Gene Accession No. 0
Gene Alias A1; CH29-493D4.3; class I MHC receptor KLRA22; killer cell lectin like receptor A1, pseudogene; killer cell lectin-like receptor 2; killer cell lectin-like receptor, subfamily A, member 1; killer cell lectin-like receptor, subfamily A, member 2; KLRA#; KLRA1; KLRA1P; Klra2; Klra22; KLRAP1; Ly49; Ly49 inhibitory receptor 8; Ly49a; ly-49a; Ly49i8; LY49L; Ly-49L; Ly49o>129>; Ly49v; lymphocyte antigen 49a; natural killer cell receptor Ly49A; novel killer cell lectin-like receptor subfamily A protein; t lymphocyte antigen A1; T-cell surface glycoprotein YE1/48
Gene Symbols KLRA1P
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human Human KLRA1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10748
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.