Learn More
ACROBiosystems ACROBiosystems Human KRAS (2-185,G12C) Protein, His Tag
Supplier:  ACROBiosystems KRSH51H3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human KRAS (2-185,G12C), His Tag (KRS-H51H3) is expressed from E.coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G12C)).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.0 kDa.
Catalog No. 50-210-6812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.