Learn More
ACROBiosystems Human KREMEN2 25ug
Supplier:  ACROBiosystems KR2H82E825UG
Encompass_Preferred
Biotinylated Human KREMEN2 Protein HisAvitag (KR2-H82E8) is expressed from human 293 cells (HEK293). It contains AA Cys 219 - Leu 326 (Accession # Q8NCW0-1).
Catalog No. 50-253-4537
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.