Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRT10 Partial ORF (NP_000412, 345 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-198 25 ug
89-950-197 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-198 Supplier Abnova™ Supplier No. H00003858Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the type I (acidic) cytokeratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Mutations in this gene are associated with epidermolytic hyperkeratosis. This gene is located within a cluster of keratin family members on chromosome 17q21. [provided by RefSeq]

Sequence: KELTTEIDNNIEQISSYKSEITELRRNVQALEIELQSQLALKQSLEASLAETEGRYCVQLSQIQAQISALEEQLQQIRAETECQNTEYQQLLDIKIRLENEIQTYRSLLE

Specifications

Accession Number NP_000412
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3858
Molecular Weight (g/mol) 37.84kDa
Name KRT10 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen KELTTEIDNNIEQISSYKSEITELRRNVQALEIELQSQLALKQSLEASLAETEGRYCVQLSQIQAQISALEEQLQQIRAETECQNTEYQQLLDIKIRLENEIQTYRSLLE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CK10/K10/KPP
Common Name KRT10
Gene Symbol KRT10
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.