Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRTAP4-4 Full-length ORF (NP_115913.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950278 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-278 25 μg
89-950-277 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-278 Supplier Abnova™ Supplier No. H00084616P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the ultrahigh sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21. [provided by RefSeq]

Sequence: MVNSCCGSVCSDQGCGLENCCRPSYCQTTCCRTTCCRPSCCVSSCCRPQCCQTTCCRTTCCHPSCCVSSCCRPQCCQSVCCQPTCCRPQCCQTTCCRTTCCRPSCCRPQCCQSVCCQPTCCCPSYCVSSCCRPQCCQTTCCRTTCCRPSCCVSRCYRPHCGQSLCC

Specifications

Accession Number NP_115913.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 84616
Molecular Weight (g/mol) 44.4kDa
Name KRTAP4-4 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MVNSCCGSVCSDQGCGLENCCRPSYCQTTCCRTTCCRPSCCVSSCCRPQCCQTTCCRTTCCHPSCCVSSCCRPQCCQSVCCQPTCCRPQCCQTTCCRTTCCRPSCCRPQCCQSVCCQPTCCCPSYCVSSCCRPQCCQTTCCRTTCCRPSCCVSRCYRPHCGQSLCC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias KAP4.13/KAP4.4/KRTAP4-13/KRTAP4.13/KRTAP4.4
Common Name KRTAP4-4
Gene Symbol KRTAP4-4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.