Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human KRTHA6 Partial ORF (NP_003762.1, 368 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950294 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-294 25 μg
89-950-293 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-294 Supplier Abnova™ Supplier No. H00008689Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq]

Sequence: DLERQNQEYQVLLDVKARLEGEIATYRHLLEGEDCKLPPQPCATACKPVIRVPSVPPVPCVPSVPCTPAPQVGTQIRTITEEIRDGKVISSREHVQSRPL

Specifications

Accession Number NP_003762.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8689
Molecular Weight (g/mol) 36.74kDa
Name KRTHA6 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen DLERQNQEYQVLLDVKARLEGEIATYRHLLEGEDCKLPPQPCATACKPVIRVPSVPPVPCVPSVPCTPAPQVGTQIRTITEEIRDGKVISSREHVQSRPL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias HA6/KRTHA6/hHa6
Common Name KRT36
Gene Symbol KRT36
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.