Learn More
Sino Biological Human KYNU PAb 100UL 10841-RP01-100
Supplier:  Sino Biological 10841RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KYNU (rh KYNU; Catalog#10841-H08B; Q16719; Met1-Asn465). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1182
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.