Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human LAG-1 (CCL4L1) Recombinant Protein, PeproTech®

Catalog No. 30058100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30058100UG 100 μg
300-58-1MG 1 mg
300-58-20UG 20 μg
30058250UG 250 μg
30058500UG 500 μg
300-58-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 30058100UG Supplier Gibco™ Supplier No. 30058100UG
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human LAG-1 is a 7.7 kDa protein containing 69 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

LAG-1 is CC chemokine that signals through the CCR5 receptor. LAG-1 is identical to MIP-1beta (ACT II isotype) except for two amino acid substitutions; arginine for histidine at position 22 and serine for glycine at position 47 of the mature protein. LAG-1 chemoattracts monocytes, and exhibits activity as an HIV-suppressive factor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NHW4
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 388372
Molecular Weight (g/mol) 7.7 kDa
Name Human LAG-1 (CCL4L1)
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name CCL4L1
Gene Symbol CCL4L1
Biological Activity Determined by its ability to chemoattract CCR5-transfected cells using a concentration of 0.1-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRSKQV CADPSESWVQ EYVYDLELN
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.