Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LAG-3 (CD223) Fc Chimera Recombinant Protein

Catalog No. PIRP88093
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88093 50 μg
PIRP88095 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88093 Supplier Invitrogen™ Supplier No. RP88093
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 1.25 mg/mL. Concentration is measured by UV.

LAG-3 is a 70-kDa surface glycoprotein in the immunoglobulin superfamily, with structural homology to CD4. It binds to MHC class II molecules with higher affinity than CD4 and is involved in the negative regulation of T cell activation and homeostatic proliferation. LAG-3 is expressed on the surface of activated T cells, including regulatory T cells, and NK cells. Notably, CD8+ T cells express LAG-3 at significantly higher levels than CD4+ T cells. Additionally, the coexpression of LAG-3 and CD49b has been proposed as a marker for identifying Type 1 regulatory T cells (Tr1 cells) in both humans and mice. This highlights LAG-3s role in immune regulation and its potential as a target for therapeutic interventions in immune-related conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18627
Concentration 0.25 mg/mL
For Use With (Application) Control, ELISA Standard, Binding Assay
Format Lyophilized
Formulation 0.1M Glutamine, 20mM sodium phosphate with 0.1M arginine, 10% glycerol, 5% mannitol, 150mM NaCl, 5% trehalose, 0.01% Tween 20, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 3902
Molecular Weight (g/mol) 84.19 kDa
Name Human LAG-3 (CD223) Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3
Common Name LAG-3 (CD223)
Gene Symbol LAG3
Biological Activity Immobilized LAG3 Protein, Human, Recombinant (ECD, hFc Tag) (at 2 μg/mL (100 μL/well) can bind Anti-LAG3 (BMS)-IgG4 Antibody (Relatlimab), the EC50 is 8-40 ng/mL)
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Human LAG3, amino acids Met1-Leu450 (Accession # NP_002277.4) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Activity Immobilized LAG3 Protein, Human, Recombinant (ECD, hFc Tag) at 2 μg/mL (100 μL/well) can bind Anti-LAG3 (BMS)-IgG4 Antibody (Relatlimab), the EC50 is 8-40 ng/mL.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.