Learn More
ACROBiosystems ACROBiosystems Human LAIR2 / CD306 Protein, His Tag
Supplier:  ACROBiosystems LA2H5220100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LAIR-2, His Tag (LA2-H5220) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Pro 152 (Accession # AAH69366).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.9 kDa. The protein migrates as 18-20 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Collagen I protein at 2 μg/mL (100 μL/well) can bind Human LAIR-2, His Tag (Cat. No. LA2-H5220) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-210-6694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.