Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LAIR2 Recombinant Protein

Catalog No. PIRP88102
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88102 1 mg
PIRP88101 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88102 Supplier Invitrogen™ Supplier No. RP88102
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (599 μL) be added to the vial to prepare a stock solution of 1.67 mg/mL. Concentration is measured by UV.

The protein encoded by this gene is a member of the immunoglobulin superfamily. It was identified by its similarity to leukocyte-associated immunoglobulin-like receptor 1, a membrane-bound receptor that modulates innate immune response. The protein encoded by this locus is a soluble receptor that may play roles in both inhibition of collagen-induced platelet aggregation and vessel formation during placental implantation. This gene maps to a region of 19q13.4, termed the leukocyte receptor cluster, which contains 29 genes in the immunoglobulin superfamily. Alternatively spliced transcript variants have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6ISS4
Concentration 0.25 mg/mL
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation 50mM tris with 5% mannitol, 0.7mM NaCl, 5% trehalose and no preservative; pH 8
Gene ID (Entrez) 3904
Molecular Weight (g/mol) 14.07 kDa
Name Human LAIR2
pH Range 8
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD306; CD306 antigen; LAIR2; LAIR-2; leukocyte associated immunoglobulin like receptor 2; leukocyte-associated Ig-like receptor 2; leukocyte-associated Ig-like receptor-2; leukocyte-associated immunoglobulin-like receptor 2; MGC71634
Common Name LAIR2
Gene Symbol LAIR2
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence Human LAIR2, amino acids Met1-Pro152 (Accession # NP_002279.2)
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥93% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.