Learn More
ACROBiosystems Human Laminin 411 Protein
Supplier:  ACROBiosystems LATH52631MG
Encompass_Preferred
Laminin 411 is a key component of the basement membrane and is involved in the regulation of cell differentiation. Laminin 411 fragment consists of the C-terminal end of the alpha 4, beta 1, and gamma 1 chains.
Catalog No. 50-253-4516
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.