Learn More
ACROBiosystems Human Laminin 521 Protein, premium grade, 1mg
Supplier:  ACROBiosystems LA5H52611MG
Encompass_Preferred
Laminin 521 (LN521) is a recombinant human protein that provides a defined surface for in vitro feeder-free culture of multiple human pluripotent stem cells (PSCs). LN521 has been proven to maintain normal growth characteristics and stemness in multiple PSC lines without simultaneous differentiation, which includes ESC, iPSC, MSC etc. In addition, LN521 has been demonstrated to support PSC growth for >10 passages without any signs of karyotypic abnormalities and to maintain the ability of PSCs to differentiate into all three germ line lineages.
Catalog No. 50-233-1755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.