Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Lass4 Synthetic Peptide

Catalog No. PIPEP12926
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPEP12926 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPEP12926 Supplier Invitrogen™ Supplier No. PEP12926
Add to Cart
Add to Cart

Synthetic Peptide

Lass4 Synthetic Peptide, PEP-12926, from Invitrogen.

The LASS (longevity assurance homolog) family members are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1, LASS2, LASS3, LASS4, LASS5 and LASS6) and they are all involved in sphingolipid synthesis. LASS4 is a 394 amino acid endoplasmic reticulum, multi-pass membrane protein. LASS4 increases the levels of long ceramides such as C22:0- and C24:0-ceramides. In cells deficient for CLN9, as observed in neuronal ceroid lipofuscinosis (NCL) or Batten disease, LASS4 can increase ceramide levels and partially correct growth and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 79603
Species Human
Purification Method purified
Accession Number Q9HA82
Common Name Human Lass4
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.08% sodium azide
For Use With (Application) Control, Western Blot
Gene Symbol CERS4
Lyophilized No
Preservative 0.08% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.