Learn More
Human LDLR Alexa Fluor 647-conjugated Antibody, R&D Systems™



Description
Specifications
Specifications
| Antigen | LDLR |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 472413 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 647 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P01130 |
| Gene Alias | FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.