Learn More
Sino Biological Human LDLR MAb 20UL 10231-MM12-20
Supplier:  Sino Biological 10231MM1220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LDLR / LDL R / LDL Receptor (rh LDLR / LDL R / LDL Receptor; Catalog#10231-H08H; NP_000518.1; Met1-Arg788). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0052
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.