Learn More
Sino Biological Human LDLR Protein 1MG 10231-H05H-1
Supplier:  Sino Biological 10231H05H1
Encompass_Preferred
A DNA sequence encoding the human LDLR (NP_000518.1) (Met1-Arg788) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-241-0170
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.