Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human LEC (CCL16) Recombinant Protein, PeproTech®

Catalog No. 30044500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30044500UG 500 μg
30044100UG 100 μg
300-44-1MG 1 mg
300-44-20UG 20 μg
30044250UG 250 μg
300-44-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 30044500UG Supplier Gibco™ Supplier No. 30044500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human LEC is an 11.2 kDa protein containing 97 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL16 gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. CCL16 displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. This cytokine also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. The expression of this gene is upregulated by IL-10.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15467
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 6360
Molecular Weight (g/mol) 11.2 kDa
Name Human LEC (CCL16)
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias C-C motif chemokine 16; C-C motif chemokine ligand 16; CCL16; chemokine (C-C motif) ligand 16; Chemokine CC-4; Chemokine LEC; CKb12; HCC-4; IL-10-inducible chemokine; ILINCK; LCC-1; LEC; liver CC chemokine-1; liver-expressed chemokine; LMC; Lymphocyte and monocyte chemoattractant; MGC117051; monotactin-1; MTN-1; NCC4; NCC-4; new CC chemokine 4; SCYA16; SCYL4; small inducible cytokine subfamily A (Cys-Cys), member 16; small-inducible cytokine A16
Common Name CCL16
Gene Symbol CCL16
Biological Activity Determined by its ability to chemoattract total human monocytes using a concentration range of 10-100 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.