Learn More
Specifications
Specifications
| Assay Sensitivity | 1.03 pg/mL |
| Sample Type | Plasma (EDTA), Heparin Plasma, Serum |
| Specificity | Natural and Recombinant Human Leptin |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3952 |
| Measuring Range | 15.60 to 1000 pg/mL |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | FLJ94114, LEP, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), OB, Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
