Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Leptin Receptor Recombinant Protein, PeproTech®

Catalog No. 30027R100UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30027R100UG 100 μg
300-27R-1MG 1 mg
30027R20UG 20 μg
30027R250UG 250 μg
30027R500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 30027R100UG Supplier Gibco™ Supplier No. 30027R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's Recombinant Human Leptin Receptor is a single, glycosylated polypeptide chain of 818 amino acid residues, corresponding to the human OB-Re isoform, and has a calculated molecular weight of approximately 93.5 kDa. As a result of glycosylation, Recombinant Human Leptin Receptor migrates with an apparent molecular mass of approximately 130-150 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The leptin (OB-R) receptor is a member of the cytokine receptor superfamily which plays an important role in mammalian body weight homeostasis and energy balance. Ligand-induced activation of the OB-R by the adipose tissue-secreted hormone, leptin, appears to activate downstream signaling events through the Janus kinase (JAK)/signal transducer and activator of transcription (STAT) pathway. Leptin can homo-dimerize OB-R extracellular domains while downstream signaling events are initiated upon homo-dimerization of OB-R intracellular domains. Several isoforms of the OB-R which differ in length and signaling capabilities have been reported. RNA transcripts encoding the OB-R long form have been found in the choroid plexus and in the hypothalamus which has been proposed as a control center for satiety and energy expenditure. Mutations in either the mouse OB-R (db) or leptin (ob) genes have been shown to result in early-onset obesity. Metabolic abnormalities attributable to these genotypes include hypercorticoidemia, hyperinsulinemia and insulin resistance, hyperglycemia, altered CNS activity, reduced metabolic rate of brown adipose tissue, and a large increase in white adipose tissue.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48357
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3953
Molecular Weight (g/mol) 93.5 kDa
Name Human Leptin Receptor
Quantity 100 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias B219; CD antigen CD295; CD295; CD295 antigen; db; diabetes; Fa; HuB219; HuB219.2; LEPR; LEP-R; Leprb; LEPRD; LEPROT; leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; Modb1; OB receptor; obese-like; obl; Obr; OB-R; OB-RGRP
Common Name Leptin Receptor
Gene Symbol Lepr
Conjugate Unconjugated
Recombinant Recombinant
Sequence FNLSYPITPW RFKLSCMPPN STYDYFLLPA GLSKNTSNSN GHYETAVEPK FNSSGTHFSN LSKTTFHCCF RSEQDRNCSL CADNIEGKTF VSTVNSLVFQ QIDANWNIQC WLKGDLKLFI CYVESLFKNL FRNYNYKVHL LYVLPEVLED SPLVPQKGSF QMVHCNCSVH ECCECLVPVP TAKLNDTLLM CLKITSGGVI FQSPLMSVQP INMVKPDPPL GLHMEITDDG NLKISWSSPP LVPFPLQYQV KYSENSTTVI READKIVSAT SLLVDSILPG SSYEVQVRGK RLDGPGIWSD WSTPRVFTTQ DVIYFPPKIL TSVGSNVSFH CIYKKENKIV PSKEIVWWMN LAEKIPQSQY DVVSDHVSKV TFFNLNETKP RGKFTYDAVY CCNEHECHHR YAELYVIDVN INISCETDGY LTKMTCRWST STIQSLAEST LQLRYHRSSL YCSDIPSIHP ISEPKDCYLQ SDGFYECIFQ PIFLLSGYTM WIRINHSLGS LDSPPTCVLP DSVVKPLPPS SVKAEITINI GLLKISWEKP VFPENNLQFQ IRYGLSGKEV QWKMYEVYDA KSKSVSLPVP DLCAVYAVQV RCKRLDGLGY WSNWSNPAYT VVMDIKVPMR GPEFWRIING DTMKKEKNVT LLWKPLMKND SLCSVQRYVI NHHTSCNGTW SEDVGNHTKF TFLWTEQAHT VTVLAINSIG ASVANFNLTF SWPMSKVNIV QSLSAYPLNS SCVIVSWILS PSDYKLMYFI IEWKNLNEDG EIKWLRISSS VKKYYIHDHF IPIEKYQFSL YPIFMEGVGK PKIINSFTQD DIEKHQSD
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.