Learn More
Invitrogen™ Human Leptin Recombinant Protein
Description
Leptin (human) Recombinant Protein, RP-8634, from Invitrogen.
Specifications
Specifications
| Accession Number | P41159 |
| Concentration | 0.1mg/mL |
| For Use With (Application) | Control |
| Formulation | 0.1%TFA with no preservative |
| Gene ID (Entrez) | 3952 |
| Molecular Weight (g/mol) | ∽16 |
| Name | Human Leptin |
| Quantity | 1 mg |
| Source | E. coli |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Show More |
in vitro research only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.