Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LHFPL5 Full-length ORF (NP_872354.1, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950627 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-627 10 μg
89-950-628 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-950-627 Supplier Abnova™ Supplier No. H00222662P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in this gene result in deafness in humans, and a mutation in a similar gene in mice results in deafness and vestibular dysfunction with severe degeneration of the organ of Corti. It is proposed to function in hair bundle morphogenesis. [provided by RefSeq]

Sequence: MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEV

Specifications

Accession Number NP_872354.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 222662
Molecular Weight (g/mol) 50.6kDa
Name LHFPL5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MVKLLPAQEAAKIYHTNYVRNSRAVGVMWGTLTICFSVLVMALFIQPYWIGDSVNTPQAGYFGLFSYCVGNVLSSELICKGGPLDFSSIPSRAFKTAMFFVALGMFLIIGSIICFSLFFICNTATVYKICAWMQLAAATGLMIGCLVYPDGWDSSEVRRMCGEQTGKYTLGHCTIRWAFMLAILSIGDALILSFLAFVLGYRQDKLLPDDYKADGTEEV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DFNB67/MGC33835/TMHS/dJ510O8.8
Common Name LHFPL5
Gene Symbol LHFPL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.