Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LIF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-5380014
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB250SP 25 μg
MAB250100 100 μg
MAB250500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB250SP Supplier R&D Systems Supplier No. MAB250SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

LIF Monoclonal specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen LIF
Applications Immunohistochemistry, Neutralization, ELISA
Classification Monoclonal
Clone 9824
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL, Neutralization 0.06-0.2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Gene Symbols LIF
Host Species Mouse
Immunogen E. coli-derived recombinant human LIF
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3976
Test Specificity Detects human LIF in ELISAs and Western blots. In sandwich immunoassays, no significant cross-reactivity or interference with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-2, rhIL-3, rhIL-4, rhIL-6, rhIL-7, rhIL-8, rhG-CSF, rhGM-CSF, rhOSM, rhTGF-beta 1, rhTNF-alpha , rhTNF-beta , recombinant mouse (rm) IL-1 beta , rmIL-3, rmIL-4, rmIL-5, rmIL-6, rmIL-7, rmGM-CSF, bovine (b) FGF acidic, bFGF basic, human (h) PDGF, porcine (p) PDGF, hTGF-beta 1, pTGF-beta 1.2, or pTGF-beta 2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.