Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LIG3 Full-length ORF (AAH09026.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89950667 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-667 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-950-667 Supplier Abnova™ Supplier No. H00003980P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is a member of the DNA ligase family. Each member of this family encodes a protein that catalyzes the joining of DNA ends but they each have a distinct role in DNA metabolism. The protein encoded by this gene is involved in excision repair and is located in both the mitochondria and nucleus, with translation initiation from the upstream start codon allowing for transport to the mitochondria and translation initiation from a downstream start codon allowing for transport to the nucleus. Additionally, alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]

Sequence: MSLAFKIFFPQTLRALSRKELCLFRKHHWRDVRQFSQWSETDLLHGHPLFLRRKPVLSFQGSHLRSRATYLVFLPGLHVGLCSGPCEMAEQRFCVDYAKRGTAGCKKCKEKIVKGVCRIGKVVPNPFSESGGDMKEWYHIKCMFEKLERARATTKKIEDLTELEGWEELEDNEKEQITQHIADLSSKAAGTPKKKAVVQAKLTTTGQVTSPVKGASFVTSTNPRKFSGFSAKPNNSGEAPSSPTPKRSLSSSKCDPRHKDCLLREFRKLCAMVADNPSYNTKTQIIQDFLRKGSAGGVA

Specifications

Accession Number AAH09026.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 3980
Molecular Weight (g/mol) 60kDa
Name LIG3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MSLAFKIFFPQTLRALSRKELCLFRKHHWRDVRQFSQWSETDLLHGHPLFLRRKPVLSFQGSHLRSRATYLVFLPGLHVGLCSGPCEMAEQRFCVDYAKRGTAGCKKCKEKIVKGVCRIGKVVPNPFSESGGDMKEWYHIKCMFEKLERARATTKKIEDLTELEGWEELEDNEKEQITQHIADLSSKAAGTPKKKAVVQAKLTTTGQVTSPVKGASFVTSTNPRKFSGFSAKPNNSGEAPSSPTPKRSLSSSKCDPRHKDCLLREFRKLCAMVADNPSYNTKTQIIQDFLRKGSAGGVA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias LIG2
Common Name LIG3
Gene Symbol LIG3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.