Learn More
ACROBiosystems ACROBiosystems Human LIGHT / TNFSF14 Protein, Mouse IgG2a Fc Tag, low endotoxin
Supplier:  ACROBiosystems LITH5256100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (LIT-H5256) is expressed from human 293 cells (HEK293). It contains AA Asp 74 - Val 240 (Accession # O43557-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 45.1 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, His Tag (Cat. No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-210-6711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.