Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA1/LILRB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30851
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30851 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB30851 Supplier R&D Systems Supplier No. MAB30851
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LILRA1/LILRB1 Monoclonal specifically detects LILRA1/LILRB1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen LILRA1/LILRB1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 586326
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75019
Gene Alias CD85I, CD85j, ILT2, LIR1, LIR6, LIR-6, MIR7
Gene Symbols LILRA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA1 Pro17-Asn461 Accession # O75019
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11024
Test Specificity Detects human LILRA1 in direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LILRB1 is observed and no cross-reactivity rhILT3, 4, 5, 6, rhLIR-7 or -8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.