Learn More
Sino Biological Human LILRA1 PAb 100UL 100479-T36-100
Supplier:  Sino Biological 100479T36100
Encompass_Preferred
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human CD85i, and purified by antigen affinity chromatography.
Catalog No. 50-240-8978
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.