Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LILRA3 (CD85e, ILT6) His-tag Recombinant Protein

Catalog No. PIRP88079
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88079 100 μg
PIRP88080 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88079 Supplier Invitrogen™ Supplier No. RP88079
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (500 μL) be added to the vial to prepare a stock solution of 2 mg/mL. Concentration is measured by UV.

Leukocyte Ig-like receptors (LIRs) are a family of immunoreceptors expressed predominantly on monocytes and B cells and at lower levels on dendritic cells and natural killer (NK) cells. All LIRs in subfamily B have an inhibitory function (see, e.g., LILRB1, MIM 604811). LIRs in subfamily A, with short cytoplasmic domains lacking an immunoreceptor tyrosine-based inhibitory motif (ITIM) and with transmembrane regions containing a charged arginine residue, may initiate stimulatory cascades (see LILRA1, MIM 604810). One member of subfamily A (LILRA3) lacks a transmembrane region and is presumed to be a soluble receptor (Borges et al., 1997 [PubMed 9548455]).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N6C8
Concentration 0.25 mg/mL
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 11026
Molecular Weight (g/mol) 57-66 kDa
Name Human LILRA3 (CD85e, ILT6) His-tag
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias CD85 antigen-like family member E; CD85e; HM31; HM43; ILT6; ILT-6; Immunoglobulin-like transcript 6; leucocyte Ig-like receptor A3; leukocyte immunoglobulin like receptor A3; leukocyte immunoglobulin-like receptor 4; leukocyte immunoglobulin-like receptor A3; leukocyte immunoglobulin-like receptor subfamily A member 3; leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member 3; LILRA3; LIR4; LIR-4; Monocyte inhibitory receptor HM43/HM31
Common Name LILRA3
Gene Symbol LILRA3
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence Human LILRA3, amino acids Met1-Glu439 (Accession # AAH28208.1) with a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >85% as determined by SDS-PAGE. >95% as determined by SEC-HPLC.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.