Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LILRA4/CD85g/ILT7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6287SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6287SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6287SP Supplier R&D Systems Supplier No. MAB6287SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT7/CD85g/LILRA4 Monoclonal specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen ILT7/CD85g/LILRA4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 656688
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P59901
Gene Alias CD85 antigen-like family member G, CD85g, CD85g antigen, ILT7, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597
Gene Symbols LILRA4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LILRA4/CD85g/ILT7 Glu24-Asn446 Accession # P59901
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23547
Test Specificity Detects human LILRA4/CD85g/ILT7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) LILRA5, rhILT2, 3, 4, 5, or 6 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.