Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LILRA4 Full-length ORF (NP_036408.3) Recombinant Protein without tag

Catalog No. 89950680 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-950-680 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-950-680 Supplier Abnova™ Supplier No. H00023547G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

This gene encodes an immunoglobulin-like cell surface protein preferentially expressed in plasmacytoid dendritic cells (PDCs). This gene is highly expressed in PDCs, and is found to be rapidly down-regulated by interleukin 3 (IL3). This gene is one of the 19 highly related genes that form a leukocyte receptor gene cluster (LRC) at chromosomal region 19q13.4. [provided by RefSeq]

Sequence: MTLILTSLLFFGLSLGPRTRVQAENLPKPILWAEPGPVITWHNPVTIWCQGTLEAQGYRLDKEGNSMSRHILKTLESENKVKLSIPSMMWEHAGRYHCYYQSPAGWSEPSDPLELVVTAYSRPTLSALPSPVVTSGVNVTLRCASRLGLGRFTLIEEGDHRLSWTLNSHQHNHGKFQALFPMGPLTFSNRGTFRCYGYENNTPYVWSEPSDPLQLLVSGVSRKPSLLTLQGPVVTPGENLTLQCGSDVGYIRYTLYKEGADGLPQRPGRQPQAGLSQANFTLSPVSRSYGGQYRCYGAHNVSSEWSAPSDPLDILIAGQISDRPSLSVQPGPTVTSGEKVTLLCQSWDPMFTFLLTKEGAAHPPLRLRSMYGAHKYQAEFPMSPVTSAHAGTYRCYGSRSSNPYLLSHPSEPLELVVSGATETLNPAQKKSDSKTAPHLQDYTVENLIRMGVAGLVLLFLGILLFEAQHSQRSPPRCSQEANSRKDNAPFRVVEPWEQI

Specifications

Accession Number NP_036408.3
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 23547
Molecular Weight (g/mol) 55.2kDa
Name LILRA4 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MTLILTSLLFFGLSLGPRTRVQAENLPKPILWAEPGPVITWHNPVTIWCQGTLEAQGYRLDKEGNSMSRHILKTLESENKVKLSIPSMMWEHAGRYHCYYQSPAGWSEPSDPLELVVTAYSRPTLSALPSPVVTSGVNVTLRCASRLGLGRFTLIEEGDHRLSWTLNSHQHNHGKFQALFPMGPLTFSNRGTFRCYGYENNTPYVWSEPSDPLQLLVSGVSRKPSLLTLQGPVVTPGENLTLQCGSDVGYIRYTLYKEGADGLPQRPGRQPQAGLSQANFTLSPVSRSYGGQYRCYGAHNVSSEWSAPSDPLDILIAGQISDRPSLSVQPGPTVTSGEKVTLLCQSWDPMFTFLLTKEGAAHPPLRLRSMYGAHKYQAEFPMSPVTSAHAGTYRCYGSRSSNPYLLSHPSEPLELVVSGATETLNPAQKKSDSKTAPHLQDYTVENLIRMGVAGLVLLFLGILLFEAQHSQRSPPRCSQEANSRKDNAPFRVVEPWEQI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CD85g/ILT7/MGC129597/MGC129598
Common Name LILRA4
Gene Symbol LILRA4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.