Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LIN-41 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5104
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5104 Supplier R&D Systems Supplier No. AF5104
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LIN-41/TRIM71 Polyclonal specifically detects LIN-41/TRIM71 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen LIN-41/TRIM71
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q2Q1W2
Gene Alias LIN41, LIN-41, TRIM71, tripartite motif containing 71, tripartite motif-containing protein 71
Gene Symbols TRIM71
Host Species Sheep
Immunogen E. coli-derived recombinant human LIN-41 Val386-Phe868 Accession # Q2Q1W2
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131405
Test Specificity Detects human LIN-41 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.