Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Lineage Cocktail 4 (lin 4) (CD2, CD3, CD4, CD7, CD8, CD10, CD11b, CD14, CD19, CD20, CD56, CD235a); BD™ Biosciences

Catalog No. BDB562722 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562722 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562722 Supplier BD Biosciences Supplier No. 562722
Add to Cart
Add to Cart

Host Species:
Clone: RPA-2.10, HIT3a, RPA-T4, M-T701, HIT8a, HI10a, ICRF44, M5E2, SJ25C1, 2H7, B159, GA-R2 (HIR2)
Isotype: Mouse IgG1, κ, Mouse IgG2a, κ, Mouse IgG2b, κ
Cross Reactivity Species: Hu

Applications:

Flow Cytometry, Fluorescence Activated Cell Sorting

TRUSTED_SUSTAINABILITY

Specifications

Classification Monoclonal
Clone RPA-2.10, HIT3a, RPA-T4, M-T701, HIT8a, HI10a, ICRF44, M5E2, SJ25C1, 2H7, B159, GA-R2 (HIR2)
Conjugate FITC
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.