Learn More
ACROBiosystems Human LIV-1 1mg
Supplier:  ACROBiosystems LV1H52571MG
Encompass_Preferred
Human LIV-1 (229-325) Protein Fc Tag (LV1-H5257) is expressed from human 293 cells (HEK293). It contains AA Lys 229 - Trp 325 (Accession # Q13433-1).
Catalog No. 50-253-4496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.