Learn More
Description
- Also known as: LOL; LOXL
- LOXL1 is a member of the lysyl oxidase family of proteins.
- It is essential to the biogenesis of connective tissue.
- It is an extracellular copper-dependent amine oxidase that catalyzes the first step in the formation of crosslinks in collagen and elastin.
- A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function.
- The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
- Mutations in LOXL1 are associated with exfoliation syndrome.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hLOXL1 |
| Accession Number | AAH68542 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1738 bp |
| Gene ID (Entrez) | 4016 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
