Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LPAR1/LPA1/EDG-2 Antibody, R&D Systems™

Catalog No. p-7235445
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB99631SP 25 μg
MAB99631100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB99631SP Supplier R&D Systems Supplier No. MAB99631SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LPAR1/LPA1/EDG-2 Monoclonal specifically detects LPAR1/LPA1/EDG-2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen LPAR1/LPA1/EDG-2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1020714
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q92633
Gene Alias edg-2, EDG2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, GPCR26, GPR26, LPA receptor 1, LPA1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, vzg-1, VZG1
Host Species Mouse
Immunogen E. coli-derived recombinant human LPAR1/LPA1/EDG-2, Met1-Lys50, Accession # Q92633
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1902
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.