Learn More
ACROBiosystems Biotinylated Human LRP-6, His,Avitag (LR6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Pro 630 (Accession # O75581-1 ).
Supplier:  ACROBiosystems LR6H82E725UG
Encompass_Preferred
LRP6 is a transmembrane low-density lipoprotein receptor that shares a similar structure with LRP5. Low-density lipoprotein receptor-related protein 6 is a key component of the LRP5/LRP6/Frizzled co-receptor group that is involved in canonical Wnt pathway. The Wnt-induced Fzd/LRP6 coreceptor complex recruits DVL1 polymers to the plasma membrane which, in turn, recruits the AXIN1/GSK3B-complex to the cell surface promoting the formation of signalsomes and inhibiting AXIN1/GSK3-mediated phosphorylation and destruction of beta-catenin.
Catalog No. 50-223-0719
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.