Learn More
ACROBiosystems Human LRRC15 2Vials
Supplier:  ACROBiosystems CHEKATP1232VIALS
Encompass_Preferred
LRRC15 (Leucine-rich repeat-containing protein 15) is also known as LIB and hLib. LRRC15 is highly expressed in a variety of solid tumors. LRRC15 was expressed on stromal fibroblasts in many solid tumors (e.g. breast head and neck lung pancreatic) as well as directly on a subset of cancer cells of mesenchymal origin (e.g. sarcoma melanoma glioblastoma). LRRC15 expression was induced by TGFβ on activated fibroblasts (αSMA+) and on mesenchymal stem cells. These collective findings suggested LRRC15 as a novel CAF and mesenchymal marker with utility as a therapeutic target for the treatment of cancers with LRRC15-positive stromal desmoplasia or cancers of mesenchymal origin. ABBV-085 is a monomethyl auristatin E (MMAE)-containing antibody–drug conjugate (ADC) directed against LRRC15 and it demonstrated robust preclinical efficacy against LRRC15 stromal-positive/cancer-negative and LRRC15 cancer-positive models as a monotherapy or in combination with standard-of-care therapies.
Catalog No. 50-253-5273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.