Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human LSAMP Full-length ORF (AAH33803, 29 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89951200 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-951-200 25 ug
89-951-199 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-951-200 Supplier Abnova™ Supplier No. H00004045P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system. During development of the limbic system, this encoded protein is found on the surface of axonal membranes and growth cones, where it acts as a selective homophilic adhesion molecule, and guides the development of specific patterns of neuronal connections. [provided by RefSeq]

Sequence: VRSVDFNRGTDNITVRQGDTAILRCVVEDKNSKVAWLNRSGIIFAGHDKWSLDPRVELEKRHSLEYSLRIQKVDVYDEGSYTCSVQTQHEPKTSQVYLIVQVPPKISNISSDVTVNEGSNVTLVCMANGRPEPVITWRHLTPTGREFEGEEEYLEILGITREQSGKYECKAANEVSSADVKQVKVTVNYPPTITESKSNEATTGRQASLKCEASAVPAPDFEWYRDDTRINSANGLEIKSTEGQSSLTVTNVTEEHYGNYTCVAANKLGVTNASLVLFRPGSVRGINGSISLAVPLWLLAASLLCLLSKC

Specifications

Accession Number AAH33803
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4045
Molecular Weight (g/mol) 59.62kDa
Name LSAMP (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 25 ug
Immunogen VRSVDFNRGTDNITVRQGDTAILRCVVEDKNSKVAWLNRSGIIFAGHDKWSLDPRVELEKRHSLEYSLRIQKVDVYDEGSYTCSVQTQHEPKTSQVYLIVQVPPKISNISSDVTVNEGSNVTLVCMANGRPEPVITWRHLTPTGREFEGEEEYLEILGITREQSGKYECKAANEVSSADVKQVKVTVNYPPTITESKSNEATTGRQASLKCEASAVPAPDFEWYRDDTRINSANGLEIKSTEGQSSLTVTNVTEEHYGNYTCVAANKLGVTNASLVLFRPGSVRGINGSISLAVPLWLLAASLLCLLSKC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias IGLON3/LAMP
Common Name LSAMP
Gene Symbol LSAMP
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.