Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human LSECtin/CLEC4G Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB2947T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB2947T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB2947T100 Supplier R&D Systems Supplier No. FAB2947T100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LSECtin/CLEC4G Monoclonal specifically detects LSECtin/CLEC4G in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LSECtin/CLEC4G
Applications Flow Cytometry
Classification Monoclonal
Clone 845404
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q6UXB4
Gene Alias CLEC4G, C-type lectin domain family 4 member G, C-type lectin domain family 4, member G, C-type lectin superfamily 4, member G, DTTR431, liver and lymph node sinusoidal endothelial cell C-type lectin, LP2698, LSECtin, Q6UXB4, UNQ431
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with LSECtin/CLEC4G Ser54-Cys293 Accession # Q6UXB4
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339390
Test Specificity Detects human LSECtin/CLEC4G in flow cytometry.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.