Learn More
ACROBiosystems Biotinylated Human LTBR, His,Avitag (LTR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Met 227 (Accession # P36941-1).
Supplier:  ACROBiosystems LTRH82E925UG
Encompass_Preferred
Lymphotoxin-beta receptor (LTBR) is also known as Tumor necrosis factor receptor superfamily member 3 (TNFRSF3), Tumor necrosis factor receptor type III (TNF-RIII), which is a single-pass type I membrane protein containing four TNFR-Cys repeat regions. Except for interacting with HCV core protein, LTBR can not only associate with itself, but also can associate with TRAF3, TRAF4 and TRAF5. As the receptor for the heterotrimeric lymphotoxin containing LTA and LTB, and for TNFS14/LIGHT, LTBR promotes apoptosis via TRAF3 and TRAF5. Furthermore, LTBR may play a role in the development of lymphoid organs.
Catalog No. 50-225-3149